Learn More
Sino Biological Human STAT6 PAb 100UL 13190-T18-100
Supplier:  Sino Biological 13190T18100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human STAT6 (rh STAT6; Catalog#13190-H08B; P42226-1; Met1-Trp847). STAT6 specific IgG was purified by Human STAT6 affinity chromatography.
Catalog No. 50-241-1986
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.