Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human STK39 Partial ORF (NP_037365, 362 a.a. - 461 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963600 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-600 10 μg
89-963-601 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-600 Supplier Abnova™ Supplier No. H00027347Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a serine/threonine kinase that is thought to function in the cellular stress response pathway. The kinase is activated in response to hypotonic stress, leading to phosphorylation of several cation-chloride-coupled cotransporters. The catalytically active kinase specifically activates the p38 MAP kinase pathway, and its interaction with p38 decreases upon cellular stress, suggesting that this kinase may serve as an intermediate in the response to cellular stress. [provided by RefSeq]

Sequence: RAKKVRRVPGSSGHLHKTEDGDWEWSDDEMDEKSEEGKAAFSQEKSRRVKEENPEIAVSASTIPEQIQSLSVHDSQGPPNANEDYREASSCAVNLVLRLR

Specifications

Accession Number NP_037365
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27347
Molecular Weight (g/mol) 36.74kDa
Name STK39 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen RAKKVRRVPGSSGHLHKTEDGDWEWSDDEMDEKSEEGKAAFSQEKSRRVKEENPEIAVSASTIPEQIQSLSVHDSQGPPNANEDYREASSCAVNLVLRLR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DCHT/DKFZp686K05124/PASK/SPAK
Common Name STK39
Gene Symbol STK39
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.