Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human STXBP5 Partial ORF (NP_640337, 1017 a.a. - 1115 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-713 10 μg
89-963-714 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-713 Supplier Abnova™ Supplier No. H00134957Q01S

Used for AP, Array, ELISA, WB-Re

Syntaxin 1 is a component of the 7S and 20S SNARE complexes which are involved in docking and fusion of synaptic vesicles with the presynaptic plasma membrane. This gene encodes a syntaxin 1 binding protein. In rat, a similar protein dissociates syntaxin 1 from the Munc18/n-Sec1/rbSec1 complex to form a 10S complex, an intermediate which can be converted to the 7S SNARE complex. Thus this protein is thought to be involved in neurotransmitter release by stimulating SNARE complex formation. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: GGGAQSLDREELFGESSSGKASRSLAQHIPGPGGIEGVKGAASGVVGELARARLALDERGQKLGDLEERTAAMLSSAESFSKHAHEIMLKYKDKKWYQF

Specifications

Accession Number NP_640337
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 134957
Molecular Weight (g/mol) 36.63kDa
Name STXBP5 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen GGGAQSLDREELFGESSSGKASRSLAQHIPGPGGIEGVKGAASGVVGELARARLALDERGQKLGDLEERTAAMLSSAESFSKHAHEIMLKYKDKKWYQF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ30922/LGL3/LLGL3/MGC141942/MGC141968/Nbla04300
Common Name STXBP5
Gene Symbol STXBP5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.