Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human STYK1 Full-length ORF (NP_060893.1, 1 a.a. - 422 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963717 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-717 10 μg
89-963-718 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-717 Supplier Abnova™ Supplier No. H00055359P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Receptor protein tyrosine kinases, like STYK1, play important roles in diverse cellular and developmental processes, such as cell proliferation, differentiation, and survival (Liu et al., 2004 [PubMed 15150103]).[supplied by OMIM]

Sequence: MGMTRMLLECSLSDKLCVIQEKQYEVIIVPTLLVTIFLILLGVILWLFIREQRTQQQRSGPQGIAPVPPPRDLSWEAGHGGNVALPLKETSVENFLGATTPALAKLQVPREQLSEVLEQICSGSCGPIFRANMNTGDPSKPKSVILKALKEPAGLHEVQDFLGRIQFHQYLGKHKNLVQLEGCCTEKLPLYMVLEDVAQGDLLGFLWTCRRDVMTMDGLLYDLTEKQVYHIGKQVLLALEFLQEKHLFHGDVAARNILMQSDLTAKLCGLGLAYEVYTRGAISSTQTIPLKWLAPERLLLRPASIRADVWSFGILLYEMVTLGAPPYPEVPPTSILEHLQRRKIMKRPSSCTHTMYSIMKSCWRWREADRPSPRELRLRLEAAIKTADDEAVLQVPELVVPELYAAVAGIRVESLFYNYSML

Specifications

Accession Number NP_060893.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55359
Molecular Weight (g/mol) 73.9kDa
Name STYK1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MGMTRMLLECSLSDKLCVIQEKQYEVIIVPTLLVTIFLILLGVILWLFIREQRTQQQRSGPQGIAPVPPPRDLSWEAGHGGNVALPLKETSVENFLGATTPALAKLQVPREQLSEVLEQICSGSCGPIFRANMNTGDPSKPKSVILKALKEPAGLHEVQDFLGRIQFHQYLGKHKNLVQLEGCCTEKLPLYMVLEDVAQGDLLGFLWTCRRDVMTMDGLLYDLTEKQVYHIGKQVLLALEFLQEKHLFHGDVAARNILMQSDLTAKLCGLGLAYEVYTRGAISSTQTIPLKWLAPERLLLRPASIRADVWSFGILLYEMVTLGAPPYPEVPPTSILEHLQRRKIMKRPSSCTHTMYSIMKSCWRWREADRPSPRELRLRLEAAIKTADDEAVLQVPELVVPELYAAVAGIRVESLFYNYSML
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp761P1010/NOK/SuRTK106
Common Name STYK1
Gene Symbol STYK1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.