Learn More
Biomatik Corporation Human Synaptosomal Associated Protein 23kDa (SNAP23) ELISA Kit
Supplier:  Biomatik Corporation EKU12542
Encompass_Preferred
Human Synaptosomal Associated Protein 23kDa (SNAP23) ELISA Kit for detecting SNAP23 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00161. Gene ID: 8773. Target Synonyms: SNAP23A; SNAP23B; Vesicle-membrane fusion protein SNAP-23. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7948
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.