Learn More
ACROBiosystems Human Syndecan-1 200tests
Supplier:  ACROBiosystems SY1HA2H9200TESTS
Encompass_Preferred
APC-Labeled Human Syndecan-1 Protein His Tag (SY1-HA2H9) is produced via conjugation of APC to Human Syndecan-1 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human Syndecan-1 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).
Catalog No. 50-253-4080
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.