Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Syntaxin 1A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7237SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7237SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7237SP Supplier R&D Systems Supplier No. MAB7237SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Syntaxin 1A Monoclonal specifically detects Syntaxin 1A in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Syntaxin 1A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 761811
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16623
Gene Alias Brain Syntaxin, HPC-1, Neuron-specific antigen HPC-1, p35-1, STX1A, STX1P35-1, SYN1A, syntaxin 1A (brain), syntaxin-1A
Gene Symbols STX1A
Host Species Mouse
Immunogen E. coli-derived recombinant human Syntaxin 1A Met1-Leu165 Accession # Q16623
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6804
Test Specificity Detects human Syntaxin 1A in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) Epimorphin/Syntaxin 2 is observed, 25% cross-reactivity with rhSyntaxin 1B2 and rhSyntaxin 3 is observed, and no cross-reactivity with rhSyntaxin 1B1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.