Learn More
Sartorius Human T Cell Killing Kit
Supplier:  Sartorius 97037
Encompass
Speed: Sample to actionable data by allowing multiple assays to be run in one assay reducing the hands on time numbers of overall tests and conserving precious sample.Results: 11 endpoints from a single well assay measuring T cell memory phenotype and function at different stages cell viability cell count and secreted pro/anti-inflammatory cytokines.Biological insight: Generate cell health and functional data from same cell by multiplexing cell and cytokine detection in one well.Simplicity: Single platform assay and data acquisition eliminates the time of acquiring and correlating data from multiple sources.
Catalog No. 50-289-1693