Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TAB3 Partial ORF (NP_690000, 58 a.a. - 149 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89963956 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-963-956 10 μg
89-963-957 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-963-956 Supplier Abnova™ Supplier No. H00257397Q01S

Used for AP, Array, ELISA, WB-Re

The product of this gene functions in the NF-kappaB signal transduction pathway. The encoded protein, and the similar and functionally redundant protein MAP3K7IP2/TAB2, forms a ternary complex with the protein kinase MAP3K7/TAK1 and either TRAF2 or TRAF6 in response to stimulation with the pro-inflammatory cytokines TNF or IL-1. Subsequent MAP3K7/TAK1 kinase activity triggers a signaling cascade leading to activation of the NF-kappaB transcription factor. The human genome contains a related pseudogene. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]

Sequence: YHSPDDNRMNRNRLLHINLGIHSPSSYHPGDGAQLNGGRTLVHSSSDGHIDPQHAAGKQLICLVQEPHSAPAVVAATPNYNPFFMNEQNRSA

Specifications

Accession Number NP_690000
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 257397
Molecular Weight (g/mol) 35.86kDa
Name TAB3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen YHSPDDNRMNRNRLLHINLGIHSPSSYHPGDGAQLNGGRTLVHSSSDGHIDPQHAAGKQLICLVQEPHSAPAVVAATPNYNPFFMNEQNRSA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC45404/NAP1/TAB3
Common Name MAP3K7IP3
Gene Symbol MAP3K7IP3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.