Learn More
ACROBiosystems ACROBiosystems Human TACI / TNFRSF13B (R119A, R122A) Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems TAIH5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human TACI (R119A, R122A), Fc Tag (TAI-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1 (R119A,R122A)).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 2 μg/mL (100 μL/well) can bind Human TACI (R119A, R122A), Fc Tag (Cat. No. TAI-H5253) with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-210-6444
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.