Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TAF2 Partial ORF (AAC02966, 504 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964004 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-004 10 ug
89-964-005 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-004 Supplier Abnova™ Supplier No. H00006873Q01S

Used for AP, Array, ELISA, WB-Re

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the larger subunits of TFIID that is stably associated with the TFIID complex. It contributes to interactions at and downstream of the transcription initiation site, interactions that help determine transcription complex response to activators. [provided by RefSeq]

Sequence: LKSISNVSGKDIQPLIKQWVDQSGVVKFYGSFAFNRKRNVLELEIKQDYTSPGTQKYVGPLKVTVQELDGSFNHTLQIEENSLKHDIPCHSKSRRNKKKK

Specifications

Accession Number AAC02966
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6873
Molecular Weight (g/mol) 36.74kDa
Name TAF2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen LKSISNVSGKDIQPLIKQWVDQSGVVKFYGSFAFNRKRNVLELEIKQDYTSPGTQKYVGPLKVTVQELDGSFNHTLQIEENSLKHDIPCHSKSRRNKKKK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CIF150/TAF2B/TAFII150
Common Name TAF2
Gene Symbol TAF2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.