Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TAF7 Partial ORF (AAH32737, 130 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964020 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-020 10 ug
89-964-021 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-020 Supplier Abnova™ Supplier No. H00006879Q01S

Used for AP, Array, ELISA, WB-Re

The intronless gene for this transcription coactivator is located between the protocadherin beta and gamma gene clusters on chromosome 5. The protein encoded by this gene is a component of the TFIID protein complex, a complex which binds to the TATA box in class II promoters and recruits RNA polymerase II and other factors. This particular subunit interacts with the largest TFIID subunit, as well as multiple transcription activators. The protein is required for transcription by promoters targeted by RNA polymerase II. [provided by RefSeq]

Sequence: FIWNHGITLPLKNVRKRRFRKTAKKKYIESPDVEKEVKRLLSTDAEAVSTRWEIIAEDETKEAENQGLDISSPGMSGHRQGHDSLEHDELREIFN

Specifications

Accession Number AAH32737
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6879
Molecular Weight (g/mol) 36.08kDa
Name TAF7 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen FIWNHGITLPLKNVRKRRFRKTAKKKYIESPDVEKEVKRLLSTDAEAVSTRWEIIAEDETKEAENQGLDISSPGMSGHRQGHDSLEHDELREIFN
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias TAF2F/TAFII55
Common Name TAF7
Gene Symbol TAF7
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.