Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TAFA-2 Recombinant Protein, PeproTech®

Catalog No. 30063250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30063250UG 250 μg
30063100UG 100 μg
300-63-1MG 1 mg
300-63-20UG 20 μg
30063500UG 500 μg
300-63-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30063250UG Supplier Gibco™ Supplier No. 30063250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TAFA-2 is an 11.2 kDa protein consisting of 101 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

This gene is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N3H0
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 338811
Molecular Weight (g/mol) 11.2 kDa
Name Human TAFA-2
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Chemokine-like protein TAFA-2; FAM19A2; family with sequence similarity 19 (chemokine (C-C motif)-like), member A2; family with sequence similarity 19 member A2, C-C motif chemokine like; protein FAM19A2; TAFA2; TAFA-2
Common Name TAFA2
Gene Symbol FAM19A2
Conjugate Unconjugated
Recombinant Recombinant
Sequence ANHHKAHHV KTGTCEVVAL HRCCNKNKIE ERSQTVKCSC FPGQVAGTTR AAPSCVDASI VEQKWWCHMQ PCLEGEECKV LPDRKGWSCS SGNKVKTTRV TH
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.