Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TAFA1 ELISA Kit

Catalog No. EH443RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH443RB 96 Tests
EH443RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH443RB Supplier Invitrogen™ Supplier No. EH443RB
Add to Cart
Add to Cart

ELISA

Human TAFA1 quantitates human TAFA1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human TAFA1.

Regulatory factor which is ligand for CMKLR2 and is involved in the modulation of neural stem-cell proliferation and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Chemokine-like protein TAFA-1; FAM19A1; family with sequence similarity 19 (chemokine (C-C motif)-like), member A1; family with sequence similarity 19 member A1, C-C motif chemokine like; protein FAM19A1; TAFA1; TAFA-1
Assay Range 0.12 to 25 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 407738
Gene Symbol FAM19A1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TAFA1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.