Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TAPBP Partial ORF (NP_003181, 197 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964062 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-062 10 ug
89-964-063 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-062 Supplier Abnova™ Supplier No. H00006892Q01S

Used for AP, Array, ELISA, WB-Re

This gene encodes a transmembrane glycoprotein which mediates interaction between newly assembled major histocompatibility complex (MHC) class I molecules and the transporter associated with antigen processing (TAP), which is required for the transport of antigenic peptides across the endoplasmic reticulum membrane. This interaction is essential for optimal peptide loading on the MHC class I molecule. Up to four complexes of MHC class I and this protein may be bound to a single TAP molecule. This protein contains a C-terminal double-lysine motif (KKKAE) known to maintain membrane proteins in the endoplasmic reticulum. This gene lies within the major histocompatibility complex on chromosome 6. Alternative splicing results in three transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: PGPPPFGLEWRRQHLGKGHLLLAATPGLNGQMPAAQEGAVAFAAWDDDEPWGPWTGNGTFWLPTVQPFQEGTYLATIHLPYLQGQVTLELAVYKPPKVSLMPAT

Specifications

Accession Number NP_003181
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6892
Molecular Weight (g/mol) 37.18kDa
Name TAPBP (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen PGPPPFGLEWRRQHLGKGHLLLAATPGLNGQMPAAQEGAVAFAAWDDDEPWGPWTGNGTFWLPTVQPFQEGTYLATIHLPYLQGQVTLELAVYKPPKVSLMPAT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias NGS17/TAPA/TPN/TPSN/tapasin
Common Name TAPBP
Gene Symbol TAPBP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.