Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TAS2R43 Full-length ORF (AAI17424.1, 1 a.a. - 309 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-095 25 μg
89-964-094 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-095 Supplier Abnova™ Supplier No. H00259289P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

TAS2R43 belongs to the large TAS2R receptor family. TAS2Rs are expressed on the surface of taste receptor cells and mediate the perception of bitterness through a G protein-coupled second messenger pathway (Conte et al., 2002 [PubMed 12584440]). For further information on TAS2Rs, see MIM 604791.[supplied by OMIM]

Sequence: MITFLPIIFSSLVVVTFVIGNFANGFIALVNSIESFKRQKISFADQILTALAVSRVGLLWVLLLNWYSTVLNPAFNSVEVRTTAYNIWAVINHFSNWLATTLSIFYLLKIANFSNFIFLHLKRRVKSVILVMLLGPLLFLACHLFVINMNEIVRTKEFEGNMTWKIKLKSAMYFSNMTVTMVANLVPFTLTLLSFMLLICSLCKHLKKMQLRGKGSQDPSTKVHIKALQTVISFLLLCAIYFLSIMISVWSFGSLENKPVFMFCKAIRFSYPSIHPFILIWGNKKLKQTFLSVFWQMRYWVKGEKTSSP

Specifications

Accession Number AAI17424.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 259289
Molecular Weight (g/mol) 61.9kDa
Name TAS2R43 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MITFLPIIFSSLVVVTFVIGNFANGFIALVNSIESFKRQKISFADQILTALAVSRVGLLWVLLLNWYSTVLNPAFNSVEVRTTAYNIWAVINHFSNWLATTLSIFYLLKIANFSNFIFLHLKRRVKSVILVMLLGPLLFLACHLFVINMNEIVRTKEFEGNMTWKIKLKSAMYFSNMTVTMVANLVPFTLTLLSFMLLICSLCKHLKKMQLRGKGSQDPSTKVHIKALQTVISFLLLCAIYFLSIMISVWSFGSLENKPVFMFCKAIRFSYPSIHPFILIWGNKKLKQTFLSVFWQMRYWVKGEKTSSP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias T2R43/T2R52
Common Name TAS2R43
Gene Symbol TAS2R43
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.