Learn More
ACROBiosystems Human Tau 2Vials
Supplier:  ACROBiosystems CHEKATP0872VIALS
Encompass_Preferred
Tau is a microtubule-associated protein which encodes by the MAPT gene that located on chromosome 17q21. Tau Promotes microtubule assembly and stability and might be involved in the establishment and maintenance of neuronal polarity. Hyperphosphorylation of the tau protein (tau inclusions pTau) can result in the self-assembly of tangles of paired helical filaments and straight filaments which are involved in the pathogenesis of Alzheimers disease frontotemporal dementia and other tauopathies.
Catalog No. 50-253-5283
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.