Learn More
ACROBiosystems Human Tau 500ug
Supplier:  ACROBiosystems TAUH5146500UG
Encompass_Preferred
Human Tau-441 K18 Pre-formed Fibrils Protein His Tag (TAU-H5146) is expressed from E. coli cells. It contains AA Gln 244 - Glu 372 (Accession # P10636-8).
Catalog No. 50-253-4074
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.