Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TCEAL2 Full-length ORF (NP_525129.1, 1 a.a. - 227 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964250 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-250 10 μg
89-964-251 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-250 Supplier Abnova™ Supplier No. H00140597P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. [provided by RefSeq]

Sequence: MEKLFNENEGMPSNQGKIDNEEQPPHEGKPEVACILEDKKLENEGNTENTGKRVEEPLKDKEKPESAGKAKGEGKSERKGKSEMQGGSKTEGKPERGGRAEGEGEPDSEREPESEGEPESETRAAGKRPAEDDIPRKAKRKTNKGLAQYLKQYKEAIHDMNFSNEDMIREFDNMARVEDKRRKSKQKLGAFLWMQRNLQDPFYPRGPREFRGGCRAPRRDTEDIPYV

Specifications

Accession Number NP_525129.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 140597
Molecular Weight (g/mol) 52.2kDa
Name TCEAL2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEKLFNENEGMPSNQGKIDNEEQPPHEGKPEVACILEDKKLENEGNTENTGKRVEEPLKDKEKPESAGKAKGEGKSERKGKSEMQGGSKTEGKPERGGRAEGEGEPDSEREPESEGEPESETRAAGKRPAEDDIPRKAKRKTNKGLAQYLKQYKEAIHDMNFSNEDMIREFDNMARVEDKRRKSKQKLGAFLWMQRNLQDPFYPRGPREFRGGCRAPRRDTEDIPYV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MY0876G05/my048
Common Name TCEAL2
Gene Symbol TCEAL2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.