Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TCEAL4 Full-length ORF (NP_001006936.1, 1 a.a. - 215 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964255 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-255 25 μg
89-964-254 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-255 Supplier Abnova™ Supplier No. H00079921P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome. Alternative splicing occurs for this gene; however, the full-length nature of all transcript variants has not yet been described. [provided by RefSeq]

Sequence: MEKLYSENEGMASNQGKMENEEQPQDERKPEVTCTLEDKKLENEGKTENKGKTGDEEMLKDKGKPESEGEAKEGKSEREGESEMEGGSEREGKPEIEGKPESEGEPGSETRAAGKRPAEDDVPRKAKRKTNKGLAHYLKEYKEAIHDMNFSNEDMIREFDNMAKVQDEKRKSKQKLGAFLWMQRNLQDPFYPRGPREFRGGCRAPRRDIEDIPYV

Specifications

Accession Number NP_001006936.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 79921
Molecular Weight (g/mol) 51kDa
Name TCEAL4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MEKLYSENEGMASNQGKMENEEQPQDERKPEVTCTLEDKKLENEGKTENKGKTGDEEMLKDKGKPESEGEAKEGKSEREGESEMEGGSEREGKPEIEGKPESEGEPGSETRAAGKRPAEDDVPRKAKRKTNKGLAHYLKEYKEAIHDMNFSNEDMIREFDNMAKVQDEKRKSKQKLGAFLWMQRNLQDPFYPRGPREFRGGCRAPRRDIEDIPYV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686M0279/FLJ21174/NPD017
Common Name TCEAL4
Gene Symbol TCEAL4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.