Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human TDP1 Recombinant Protein
SDP

Catalog No. NB5966150UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB5966150UG 50 μg
NB5966110UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB5966150UG Supplier Novus Biologicals™ Supplier No. NBP25966150UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-608 of Human TDP1 The Recombinant Human TDP1 Protein is derived from E. coli. The Recombinant Human TDP1 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) PAGE
Formulation In PBS (pH 7.4) containing 10% glycerol
Molecular Weight (g/mol) 69.5 kDa
Name TDP1 Recombinant Protein
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias FLJ13946, FLJ77601
Gene Symbol TDP1
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.