Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Tenascin X Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6999
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6999 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6999 Supplier R&D Systems Supplier No. AF6999
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Tenascin X Polyclonal specifically detects Tenascin X in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Tenascin X
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. AAH33740
Gene Alias Hexabrachion-like protein, HXBL, tenascin XB, tenascin XB2, tenascin-X, TNX, TNXB, TNXB1growth-inhibiting protein 45, TNXB2tenascin XB1, TNXBS, TNXBSTNXTENX, XB, XBS, XB-S, XBTN-X
Gene Symbols TNXB
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Tenascin X isoform 2 Met1-Gly673 Accession # AAH33740
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7148
Test Specificity Detects recombinant human Tenascin X isoform 2 in direct ELISAs and human Tenascin X in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Tenascin R is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.