Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TEP1 Partial ORF (NP_009041, 2529 a.a. - 2627 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964391 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-391 10 ug
89-964-392 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-391 Supplier Abnova™ Supplier No. H00007011Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene product is a component of the ribonucleoprotein complex responsible for telomerase activity which catalyzes the addition of new telomeres on the chromosome ends. The telomerase-associated proteins are conserved from ciliates to humans. [provided by RefSeq]

Sequence: ASMDSDASMDSEPTPHLKTRQRRKIHSGSVTALHVLPELLVTASKDRDVKLWERPSMQLLGLFRCEGSVSCLEPWLGANSTLQLAVGDVQGNVYFLNWE

Specifications

Accession Number NP_009041
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7011
Molecular Weight (g/mol) 36.63kDa
Name TEP1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen ASMDSDASMDSEPTPHLKTRQRRKIHSGSVTALHVLPELLVTASKDRDVKLWERPSMQLLGLFRCEGSVSCLEPWLGANSTLQLAVGDVQGNVYFLNWE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias TLP1/TP1/TROVE1/VAULT2/p240
Common Name TEP1
Gene Symbol TEP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.