Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TEX19 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6319
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6319 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6319 Supplier R&D Systems Supplier No. AF6319
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

TEX19 Polyclonal specifically detects TEX19 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen TEX19
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunocytochemistry 10 ug/mL
Gene Accession No. Q8NA77
Gene Alias FLJ35767, testis expressed 19, testis-expressed sequence 19 protein
Gene Symbols TEX19
Host Species Sheep
Immunogen E. coli-derived recombinant human TEX19 Met1-Ser164 Accession # Q8NA77
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400629
Test Specificity Detects human TEX19 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human TEX11 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.