Learn More
Sino Biological Human TF PAb 100UL 11019-RP02-100
Supplier:  Sino Biological 11019RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Transferrin / TF (rh Transferrin; Catalog#11019-H08H; NP_001054.1; Met 1-Pro 698). Transferrin specific IgG was purified by human Transferrin affinity chromatography.
Catalog No. 50-240-9213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.