Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TFF-3 Recombinant Protein, PeproTech®

Catalog No. 300611MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-61-1MG 1 mg
30061100UG 100 μg
300-61-20UG 20 μg
30061250UG 250 μg
30061500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 300-61-1MG Supplier Gibco™ Supplier No. 300611MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TFF-3 is a 13.2 kDa homodimeric protein consisting of two 59 amino acid chains, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. Trefoil peptides contain three intrachain disulfide bonds, forming the trefoil motif, or P-domain and clustered in human chromosome 21q22.3. Trefoil Factor 3 (TFF3) is one of three trefoil peptides secreted by epithelial cells that line mucus membranes in the large and small intestines and lower respiratory tract. TFF3 is mainly active as a disulfide-linked homodimer and may have a role in promoting cell migration in healing processes. It is involved in the maintenance and repair of the intestinal mucosa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07654
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7033
Molecular Weight (g/mol) 13.2 kDa
Name Human TFF-3
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias hITF; hP1.B; Intestinal trefoil factor; ITF; mITF; P1B; Polypeptide P1.B; rITF; rP1.B; Tff3; TFI; Trefoil factor 3; trefoil factor 3 (intestinal); trefoil factor 3, intestinal
Common Name TFF3
Gene Symbol TFF3
Biological Activity Determined by its ability to chemoattract human MCF-7 cells using a concentration range of 1.0-10.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence EEYVGLSANQ CAVPAKDRVD CGYPHVTPKE CNNRGCCFDS RIPGVPWCFK PLQEAECTF
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.