Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human TGF-Beta 1, Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 100395UG
Encompass_Preferred
Transforming growth factor-beta 1 (TGF-B 1) is a member of the TGF-beta superfamily of cytokines. Members of this family exhibit regulatory activity in immunity, proliferation, adhesion, migration, and growth inhibition pathways. TGF-B 1signals through SMAD proteins via the TGF-bRI and TGF-bRII receptors.
Catalog No. 50-589-860
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.