Learn More
Gibco™ Human TGF-beta 1 Recombinant Protein, PeproTech®
Description
Recombinant Human TGF-beta1 is a 25.0 kDa homo-dimer, consisting of two identical 112 amino acid polypeptide chains, each weighing 12.5 kDa and linked by a single disulfide bond. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P01137 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 7040 |
| Molecular Weight (g/mol) | 25 kDa |
| Name | Human TGF-beta 1 |
| Purification Method | Purified |
| Quantity | 250 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.