Learn More
ACROBiosystems Human TGF-beta RI 1mg
Supplier:  ACROBiosystems TG1H52541MG
Encompass_Preferred
Human TGF-beta RI Protein Fc Tag (TG1-H5254) is expressed from human 293 cells (HEK293). It contains AA Leu 34 - Leu 126 (Accession # P36897-1).
Catalog No. 50-253-4063
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.