Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Human Th17 Cytokine Staining Panel

Catalog No. 5017497
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 panel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-174-97 1 panel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-174-97 Supplier Invitrogen™ Supplier No. 88841944
  • $1,192.00
    $893.65 / Each of 1

    Save $298.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Anti-Human IL-17A (eBio64Dec17) FITC : 25 tests (0.125 μg/5 μL test)*
  • Anti-Human IL-17F (SHLR17) PE : 25 tests (0.5 μg/5 μL test)*
  • Anti-Human IL-22 (22URTI) PerCP-eFluor 710 : 25 tests (0.015 μg/5 μL test)*
  • Anti-Human IL-21 (3A3-N2) eFluor 660 : 25 tests (0.125 μg/5 μL test)*
  • Anti-Human CD4 (RPA-T4) eFluor 450 : 25 tests (0.25 μg/5 μL test)*
  • IC Fixation Buffer
  • Permeabilization Buffer (10x)
  • *A test is defined as the amount of antibody to use for 1 million cells in a 100 μL staining volume.

Includes all reagents needed for simultaneous flow cytometric detection of all the major cytokines produced by the Th17 lineage (IL-17A, IL-17F, IL-21 and IL-22).

  • 5μL/test
  • Also included: An anti-CD4 antibody that can be used after fixation and permeabilization, as well as IC Fixation and Permeabilization Buffers
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C. Light sensitive material.
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 1 panel
For Use With (Application) Intracellular Staining, Flow Cytometry
Product Line eBioscience
Product Type Staining Panel

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.