Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Thioredoxin-like 5/TRP14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3504
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3504 Supplier R&D Systems Supplier No. MAB3504
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Thioredoxin-like 5/TRP14/TXNDC17 Monoclonal specifically detects Thioredoxin-like 5/TRP14/TXNDC17 in Human samples. It is validated for Western Blot.

Specifications

Antigen Thioredoxin-like 5/TRP14/TXNDC17
Applications Western Blot
Classification Monoclonal
Clone 544710
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BRA2
Gene Alias MGC14353, Protein 42-9-9, thioredoxin (Trx)-related protein, 14 kDa, thioredoxin domain containing 17, thioredoxin domain-containing protein 17, Thioredoxin like 5, thioredoxin-like 5, Thioredoxin-like protein 5, TRP14, TRP1414 kDa thioredoxin-related protein, TXNDC17, TXNL5
Gene Symbols TXNDC17
Host Species Mouse
Immunogen E. coli-derived recombinant human TRP14 Ala2-Asp123 Accession # Q9BRA2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84817
Test Specificity Detects endogenous human TRP14 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.