Learn More
ILEX LIFE SCIENCES LLC HUMAN THROMBIN PROTEIN 1 MG
SDP
Supplier:  ILEX LIFE SCIENCES LLC A13391000
Encompass_Preferred
Ilex Life Sciences Thrombin Human, Purified Protein (Plasma), 1 mg. CAS REGISTRY NO.: 9002-04-4. BIOLOGICAL ACTIVITY: The specific activity was found to be 2435 NIH Units/mg (NOTE: may vary with each lot number, please refer to the datasheet enclosed with your product). FORMULATION: Sterile filtered liquid solution; contains 0.15M NaCl, 0.1M Tris, pH 7.4. APPLICATION: As a result of its high proteolytic specificity, thrombin has become an important biochemical protein. The thrombin cleavage site (Leu-Val-Pro-Arg-Gly-Ser) is widely used in linker regions of recombinant fusion protein constructs. After the purification of the fusion protein, thrombin is used to cleave between the Arginine and Glycine residues of the cleavage site, efficiently removing the purification tag from the protein of interest with a high degree of specificity. OTHER NAMES: Native Activated Factor IIa, Factor 2A, F2, PT, RPRGL2, THPH1, coagulation factor II, EC 3.4.21.5, UniProtKB# P00734.
Catalog No. 50-273-4022
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.