Learn More
ACROBiosystems Human Thrombopoietin / TPO Protein, premium grade
Supplier:  ACROBiosystems THNH521620UG
Encompass_Preferred
Thrombopoietin (TPO) is a 332 amino acid glycoprotein made primarily in the liver and act as the major physiological regulator of platelet production. TPO binds the TPO receptor, activates JAK and STAT pathways, thus stimulating megakaryocyte growth and platelet production. Additionally, further investigation uncovered that thrombopoietin is a critical cytokine promoting hematopoietic rebound after myeloablation and its transcripts are expressed by multiple cellular sources.
Catalog No. 50-271-8828
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.