Learn More
R&D Systems™ Human Thrombospondin-1 Quantikine ELISA Kit
Description
Specifications
Specifications
| Assay Range | 7.81 to 500 ng/mL |
| Assay Sensitivity | 0.944 ng/mL |
| Conjugate | HRP |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Human Milk, Platelet-poor Plasma (EDTA), Platelet-poor Plasma (EDTA, Heparin), Platelet-poor Plasma (Heparin), Saliva, Serum |
| Specificity | Natural and Recombinant Human Thrombospondin-1 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7057 |
| Gene Symbol | THBS1 |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.