Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Human Thymosin Beta 4 Synthetic Peptide, >95% Purity, 100 μg
Description
- Peptide Name: Thymosin beta 4
- Type: Synthetic
- Applications: BL
- Purity: >95%
- Form: Lyophilized
- Storage Buffer: 0.1% Acetic acid
- Species Reactivity: Human
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Thymosin Beta 4 (Human) |
| Substrate Properties | Peptide-based Substrate |
| Reactivity | Human |
| Shipping Condition | Blue Ice |
| Label or Dye | Unconjugated |
| Content And Storage | Store at -20°C. |
| Product Type | Blocker and Diluent |
| Buffer | 0.1% acetic acid |
| Form | Lyophilized |
| Formulation | Lyophilized from 0.1% Acetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?