Learn More
American Research Products Inc Human thyroXine T4 ELISA Kit
Supplier:  American Research Products Inc CSBE05081H
Encompass
Human thyroXine T4 ELISA Kit from CUSABIO Detection Range 20 ng/mL-320 ng/mL Sensitivity 20 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29405