Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TICAM1 Partial ORF (-, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89965632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-965-632 25 μg
89-965-631 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-965-632 Supplier Abnova™ Supplier No. H00148022Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

TRIF, or TICAM1, is a Toll/IL1R (MIM 147810) (TIR) domain-containing adaptor molecule, like MYD88 (MIM 602170) and TIRAP (MIM 606252), that induces interferon-beta (IFNB; MIM 147640), specifically interacts with TLR3 (MIM 603029), and activates nuclear factor kappa-B (NFKB; see MIM 164011).[supplied by OMIM]

Sequence: AIDHSAFIILLLTSNFDCRLSLHQVNQAMMSNLTRQGSPDCVIPFLPLESSPAQLSSDTASLLSGLVRLDEHSQIFARKVANTFKPHRLQARKAMWRKEQDTRALREQSQ

Specifications

For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 148022
Molecular Weight (g/mol) 37.84kDa
Name TICAM1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen AIDHSAFIILLLTSNFDCRLSLHQVNQAMMSNLTRQGSPDCVIPFLPLESSPAQLSSDTASLLSGLVRLDEHSQIFARKVANTFKPHRLQARKAMWRKEQDTRALREQSQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC35334/PRVTIRB/TICAM-1/TRIF
Common Name TICAM1
Gene Symbol TICAM1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.