Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TIGAR-TAT Recombinant Protein, PeproTech®

Catalog No. 15014T250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
250 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15014T250UG 250 μg
15014T100UG 100 μg
15014T25UG 25 μg
15014T500UG 500 μg
150-14T-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 15014T250UG Supplier Gibco™ Supplier No. 15014T250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TIGAR-TAT expressed in E.coli is a 31.6 kDa protein containing 283 amino acid residues, including the 269 residues of full-length TIGAR fused to a 14-residue C-terminal peptide containing the TAT transduction domain (GGGYGRKKRRQRRR). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TIGAR is a p53-inducible enzyme that catalyzes the hydrolysis of fructose-2-6 bisphosphate (F-2-6-BP) to fructose-6-phosphate and inorganic phosphate. F-2-6-BP is a powerful activator of 6-phosphofructose-1 kinase, the rate limiting enzyme of glycolysis. By lowering the intracellular level of F-2-6-BP, TIGAR expression leads to increased glucose processing via the pentose phosphate pathway, the major cellular source for NADPH. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing transcription factors and other intracellular proteins into primary, as well as transformed, cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NQ88
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 57103
Molecular Weight (g/mol) 31.6 kDa
Name Human TIGAR-TAT
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias TIGAR
Common Name TIGAR-TAT
Gene Symbol TIGAR
Biological Activity Pretreatment with TIGAR-TAT for 4 hrs, using a concentration range 0.1-5.0 ug/ml, protects U2OS cells from apoptosis induced by hydrogen peroxide.
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag TAT-fusion
Sequence ARFALTVVRH GETRFNKEKI IQGQGVDEPL SETGFKQAAA AGIFLNNVKF THAFSSDLMR TKQTMHGILE RSKFCKDMTV KYDSRLRERK YGVVEGKALS ELRAMAKAAR EECPVFTPPG GETLDQVKMR GIDFFEFLCQ LILKEADQKE QFSQGSPSNC LETSLAEIFP LGKNHSSKVN SDSGIPGLAA SVLVVSHGAY MRSLFDYFLT DLKCSLPATL SRSELMSVTP NTGMSLFIIN FEEGREVKPT VQCICMNLQD HLNGLTETRG GGYGRKKRRQ RRR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.