Learn More
Invitrogen™ Human TIMP-1 ELISA (For Lysates) Kit
Description
Human TIMP-1 quantitates human TIMP-1 in cell lysate, tissue homogenate. The assay will exclusively recognize both natural and recombinant human TIMP-1.
Specifications
Specifications
| Accession Number | Clgi; Collagenase inhibitor; collagenase inhibitor 16C8 fibroblast; EG-1; embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; LOC100009047; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; TIMP; TIMP 1; TIMP metallopeptidase inhibitor 1; Timp1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1 |
| Assay Range | 30 to 18,000 pg/mL |
| Assay Sensitivity | 30 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Cell Lysates, Tissue Homogenates |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 7076 |
| Gene Symbol | TIMP1 |
| Interassay CV | <12% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.