Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human TIMP-2 Recombinant Protein, PeproTech®

Catalog No. 41002250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41002250UG 250 μg
41002100UG 100 μg
410-02-10UG 10 μg
410-02-1MG 1 mg
410-02-2UG 2 μg
41002500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 41002250UG Supplier Gibco™ Supplier No. 41002250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human TIMP-2 is a 21.8 kDa protein containing 194 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P16035
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7077
Molecular Weight (g/mol) 21.8 kDa
Name Human TIMP-2
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Collagenase inhibitor; CSC-21 K; D11Bwg1104e; DDC8; etID32613.12; fa96h09; Metalloproteinase inhibitor 2; metalloproteinase inhibitor 2 A; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; metalloproteinase-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; TIMP metallopeptidase inhibitor 2 A; TIMP2; Timp-2; TIMP-2 protein; timp2a; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase 2 precursor; tissue inhibitor of metalloproteinase 2 A; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinase-2 precursor; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2; wu:fa96h09; zgc:136726
Common Name TIMP2
Gene Symbol TIMP2
Biological Activity TIMP-2 activity was measured by its ability to inhibit human MMP-1-induced hydrolysis of a chromogenic peptide substrate at room temperature. Half maximal inhibition was obtained (at a TIMP-2 concentration of approximately 0.5 ug/ml, when using an MMP-1 concentration of 1.6 ug/ml)
Conjugate Unconjugated
Recombinant Recombinant
Sequence CSCSPVHPQQ AFCNADVVIR AKAVSEKEVD SGNDIYGNPI KRIQYEIKQI KMFKGPEKDI EFIYTAPSSA VCGVSLDVGG KKEYLIAGKA EGDGKMHITL CDFIVPWDTL STTQKKSLNH RYQMGCECKI TRCPMIPCYI SSPDECLWMD WVTEKNINGH QAKFFACIKR SDGSCAWYRG AAPPKQEFLD IEDP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.