Learn More
R&D Systems™ Human TIMP Luminex™ Performance Assay 3-plex Fixed Panel
Shop All Novus Biologicals Products

Description
- Magnetic format allows for easier wash steps
- Undergoes similar validation testing as Quantikine™ ELISA Kits
- Precise and Specific
- Requires a small sample volume (<50 μL)
- Run your assay in just 3-5 hours
- Mass-calibrated standards for consistent results with every new lot of material
- Enough reagents for one 96-well plate
Specifications
Specifications
| Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
| Quantity | 1 Kit |
| Species | Human |
| Binding Target | TIMP Assay Kits |
| Bead Type | Magnetic |
| Gene ID (Entrez) | 7076 |
| Product Line | R&D Systems |
| Sample Type | Serum, Urine, Saliva, Plasma (EDTA, Heparin), Cell Culture Supernatant |
| Sample Volume | Cell Culture Supernates (10μL),Serum (10μL),EDTA Plasma (10μL),Heparin Plasma (10μL),Saliva (10μL),Urine (10μL),Human Milk (10μL) |
| Kit Contents |
|
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.