Learn More
Biomatik Corporation Human Tissue Factor (TF) CLIA Kit
Supplier:  Biomatik Corporation EKU09465
Encompass_Preferred
Human Tissue Factor (TF) CLIA Kit for detecting TF in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.97pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P13726. Gene ID: 2152. Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8513
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.