Learn More
Creative Biomart Anistreplase; Human tissue plasminogen activator, tPA
Supplier:  Creative Biomart THP0011
Encompass_Preferred
Human tissue plasminogen activator, purified, glycosylated, 527 residues purified from CHO cells. It is the p-anisoyl derivative of the primary Lys-plasminogen-streptokinase activator complex (a complex of Lys-plasminogen and streptokinase). A p-anisoyl group is chemically conjugated to a complex of bacterial-derived streptokinase and human Plasma-derived Lys-plasminogen proteins.
Catalog No. 50-208-4062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.