Learn More
ACROBiosystems Human TL1A Protein His Tag
Supplier:  ACROBiosystems TLAH52451MG
Encompass_Preferred
Human TL1A Protein His Tag (TLA-H5245) is expressed from human 293 cells (HEK293). It contains AA Pro 23 - Leu 174 (Accession # O95150-3).
Catalog No. 50-253-4041
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.