Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human TL1A/TNFSF15 DuoSet ELISA
SDP

Catalog No. DY131905 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
Five 96-well plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DY131905 Five 96-well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY131905 Supplier R&D Systems™ Supplier No. DY131905
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

For determination of natural and recombinant Interleukin human Tumor Necrosis Factor-like proteins in humans.

The Human TL1A/TNFSF15 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TL1A/TNFSF15.

Specifications

Assay Range 18.80 to 1200pg/mL
Immunoassay Kit Format 96-well Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sufficient For Five 96-well plates
Target Species Human
Gene ID (Entrez) 9966 (Human); 326623 (Mouse);
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target TL1A/TNFSF15

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.