Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TLN1 Partial ORF (AAH42923.1, 2293 a.a. - 2533 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89964856 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-964-856 25 ug
89-964-855 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-964-856 Supplier Abnova™ Supplier No. H00007094Q08L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a cytoskeletal protein that is concentrated in areas of cell-substratum and cell-cell contacts. The encoded protein plays a significant role in the assembly of actin filaments and in spreading and migration of various cell types, including fibroblasts and osteoclasts. It codistributes with integrins in the cell surface membrane in order to assist in the attachment of adherent cells to extracellular matrices and of lymphocytes to other cells. The N-terminus of this protein contains elements for localization to cell-extracellular matrix junctions. The C-terminus contains binding sites for proteins such as beta-1-integrin, actin, and vinculin. [provided by RefSeq]

Sequence: TEWVDPEDPTVIAENELLGAAAAIEAAAKKLEQLKPRAKPKEADESLNFEEQILEAAKSIAAATSALVKAASAAQRELVAQGKVGAIPANALDDGQWSQGLISAARMVAAATNNLCEAANAAVQGHASQEKLISSAKQVAASTAQLLVACKVKADQDSEAMKRLQAAGNAVKRASDNLVKAAQKAAAFEEQENETVVVKEKMVGGIAQIIAAQEEMLRKERELEEARKKLAQIRQQQYKFL

Specifications

Accession Number AAH42923.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7094
Molecular Weight (g/mol) 52.14kDa
Name TLN1 (Human) Recombinant Protein (Q08)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 25 ug
Immunogen TEWVDPEDPTVIAENELLGAAAAIEAAAKKLEQLKPRAKPKEADESLNFEEQILEAAKSIAAATSALVKAASAAQRELVAQGKVGAIPANALDDGQWSQGLISAARMVAAATNNLCEAANAAVQGHASQEKLISSAKQVAASTAQLLVACKVKADQDSEAMKRLQAAGNAVKRASDNLVKAAQKAAAFEEQENETVVVKEKMVGGIAQIIAAQEEMLRKERELEEARKKLAQIRQQQYKFL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ILWEQ/KIAA1027/TLN
Common Name TLN1
Gene Symbol TLN1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.