Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TMEM18 Synthetic Peptide

Catalog No. PEP0804
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0804 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0804 Supplier Invitrogen™ Supplier No. PEP0804
Add to Cart
Add to Cart

Synthetic Peptide

TMEM18 Synthetic Peptide, PEP-0804, from Invitrogen.

Recent studies have examined how overexpression of certain genes enhances the tropism of neural stem cells (NSCs) for gliomas. Overexpression of Transmembrane protein 18 (TMEM18) provides NSCs and neural precursors an increased migration capacity towards glioblastoma cells in vitro and in the rat brain, while functional inactivation of the gene resulted in an almost complete loss of migration activity, suggesting that TMEM18 could be a potential target in NSC-based glioma therapy. Other recent results suggest that among others, the TMEM18 loci is associated with higher than normal body mass index (BMI) and obesity, indicating there may be a CNS-related role in predisposition to obesity. Multiple isoforms of TMEM18 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 129787
Species Human
Purification Method purified
Accession Number Q96B42
Common Name Human TMEM18
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol TMEM18
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.