Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human TNF (127 a.a.) Full-lenth Recombinant Protein expressed in Barley grain (Hordeum vulgare)

Catalog No. 89940699 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-940-699 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-940-699 Supplier Abnova™ Supplier No. P5885
Add to Cart
Add to Cart

Used for Func, SDS-PAGE, WB-Re

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes and macrophages. The active form of the protein is found extracellularly as a homodimer. This gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. [provided by RefSeq]

Specifications

For Use With (Application) Functional Assay, SDS-PAGE, Western Blot
Formulation Lyophilized from PBS, pH 7.2
Gene ID (Entrez) 1437
Molecular Weight (g/mol) 30, 38kDa
Name Human TNF (127 a.a.) Full-lenth Recombinant Protein expressed in Barley grain (Hordeum vulgare)
Purification Method Chromatography
Quality Control Testing SDS-PAGE Stained with Coomassie Blue
Quantity 1 mg
Storage Requirements Store at −20°C.Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.005ng/μg (0.05EU/μg) as measured by LAL assay.
Gene Alias GMCSF, MGC131935, MGC138897
Common Name CSF2
Gene Symbol CSF2
Biological Activity Activity has been measured by cell-based proliferation assay using TF-1 cells. ED50 is 0.05ng/mL.
Species Plants
Recombinant Recombinant
Protein Tag Untagged
Expression System Hordeum vulgare (barley) expression system
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.