Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human TNF-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7067879
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2101RSP 25 μg
MAB2101R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2101RSP Supplier R&D Systems Supplier No. MAB2101RSP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TNF-alpha Monoclonal specifically detects TNF-alpha in Human samples. It is validated for Neutralization, Immunocytochemistry.

Specifications

Antigen TNF-alpha
Applications Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 6401R
Conjugate Unconjugated
Dilution Neutralization 0.1-0.6 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01375
Gene Alias APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Gene Symbols TNF
Host Species Mouse
Immunogen E. coli-derived recombinant human TNF-alpha Gly57-Leu233 Accession # P01375
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7124
Test Specificity Detects human TNF-alpha in direct ELISAs and Western blots. In direct ELISAs, approximately 25-50% cross-reactivity with recombinant porcine TNF-alpha and recombinant rhesus macaque TNF-alpha is observed but no cross-reactivity with recombinant cotton rat TNF-alpha , recombinant rat TNF-alpha , recombinant human (rh) LT alpha 1/beta 2, rhLT alpha 2/beta 1, rhAPRIL, rhBAFF, rhEDA-A2, recombinant mouse EDA, rhFas Ligand, rhLIGHT, rhOX40 Ligand, rhTRAIL, rhTRANCE, rhTWEAK, or rhVEGI is observed.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.