Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human TNF-alpha Quantikine ELISA Kit
SDP

Catalog No. STA00D Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Pack (6 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STA00D 1 Pack (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. STA00D Supplier R&D Systems™ Supplier No. STA00D
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TNF-alpha Quantikine ELISA Kit quantifies human TNF-alpha in cell culture supernates (50 ul), serum (50 ul), plasma (50 ul).

1 Pack (6 Plates)

Specifications

Assay Range 15.60 - 1000 pg/mL
Assay Sensitivity 6.23 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity HIV-1 Gag p24
For Use With (Application) ELISA
Gene ID (Entrez) 7124
Host Species Human
Quantity 1 Pack (6 Plates)
Sample Volume Cell Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Target TNF-alpha
Total Assay Time 4.0 hours
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.